Learn More
CD97 Rat anti-Mouse, Alexa Fluor™ 350, Clone: 2Q42, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222000AF350
Catalog No. NP22200A350
Add to cart
Description
Description
CD97 Monoclonal antibody specifically detects CD97 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD97
2Q42
Flow Cytometry
CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1
Rat
IgG1
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD97
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein CD97. Rat monoclonal anti mouse CD97 AAH06676.
0.1 mL
Primary
976