Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD97 Antibody (2Q42), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP222000AF750
Description
CD97 Monoclonal specifically detects CD97 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD97 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ADGRE5 | |
Mouse recombinant CD97 | |
Primary | |
Mouse | |
IgG1 |
Flow Cytometry | |
2Q42 | |
Flow Cytometry | |
CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
Rat | |
0.1 mL | |
976 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction