Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD98 Antibody (UM7F8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234506AF532
Description
CD98 Monoclonal specifically detects CD98 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD98 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SLC3A2 | |
Molt 13 T cell line | |
0.1 mL | |
6520 | |
Human | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
UM7F8 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen | |
Mouse | |
Protein A or G purified | |
Primary | |
CD98 exits as a heterodimer containing a disulphide-linked glycosylated heavy chain and a non-glycosylated light chain. It is a member of the solute carrier family and encodes a cell surface, transmembrane protein. The protein exists as the heavy chain of a heterodimer, covalently bound through disulfide bonds to one of several possible light chains. The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids. Alternatively spliced transcript variants, encoding different isoforms, have been characterized. | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction