Learn More
CD98 Mouse anti-Human, FITC, Clone: MEM-108, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15739
Catalog No. 50-113-7088
Add to cart
Description
Description
CD98 (4F2) is a type II transmembrane glycoprotein which serves as the heavy chain of the heterodimeric amino acid transporters (HATs). CD98, linked to various light chains by disulfide bond, is responsible for cell surface expression and basolateral localization of this transporter complex in polarized epithelial cells and also interacts with beta1 integrins and increases their affinity for ligand. Besides its roles in amino acid transport, CD98 is thus involved in cell fusion and activation. It is implicated in regulation of cellular differentiation, growth and apoptosis.
Specifications
Specifications
CD98
Monoclonal
FITC
SLC3A2
Liquid
SLC3A2
Human CD98
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MEM-108
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P08195
4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE
Mouse
IgG1
100 tests
4° C
Primary
6520