Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99 Mouse anti-Human, Clone: HO36-1.1, Abnova™

Mouse monoclonal antibody raised against native human CD99.

Manufacturer:  Abnova Corporation MAB13349

Catalog No. 89-933-892


Add to cart

Description

Description

The protein encoded by this gene is a cell surface glycoprotein involved in leukocyte migration, T-cell adhesion, ganglioside GM1 and transmembrane protein transport, and T-cell death by a caspase-independent pathway. In addition, the encoded protein may have the ability to rearrange the actin cytoskeleton and may also act as an oncosuppressor in osteosarcoma. Cyclophilin A binds to CD99 and may act as a signaling regulator of CD99. This gene is found in the pseudoautosomal region of chromosomes X and Y and escapes X-chromosome inactivation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

CD99
HO36-1.1
Mouse monoclonal antibody raised against native human CD99.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
MIC2/MIC2X/MIC2Y
Mouse
IgM, kappa
100 ug
Store at 4°C.
Monoclonal
This monoclonal antibody shows a very similar reactivity to other CD99 monoclonal antibodies.
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (5-10 uL/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CD99
Purified E-rosette forming cells from human peripheral blood lymphocytes.
Protein A/G purification
RUO
Primary
4267
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit