Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC123 Mouse anti-Human, Canine, Monkey, Alexa Fluor™ 700, Clone: OTI1F8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271391AF700

Catalog No. N271391A700


Add to cart

Description

Description

CDC123 Monoclonal antibody specifically detects CDC123 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

CDC123
OTI1F8
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
CDC123
Full length recombinant protein of human CDC123 (NP_006014) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication
Primary
8872.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
cell division cycle 123 homolog (S. cerevisiae), cell division cycle protein 123 homolog, chromosome 10 open reading frame 7, D123C10orf7, FLJ13863, HT-1080, Protein D123, PZ32
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit