Learn More
Abnova™ CDC14B 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00008555T01
Catalog No. 89-938-841
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CDC14B 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 51.92kDa
Transient overexpression cell lysate was tested with anti-CDC14B antibody (Mfr. No. H00008555-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
ENST00000265659
pCMV-CDC14B full-length
100 μL
Human