Learn More
Alfa Aesar™ Cdc2-Like Kinase Inhibitor, TG003
Manufacturer:  Alfa Aesar™ J64690LB0
 View more versions of this product
Catalog No. AAJ64690LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members.
Specifications
Specifications
5mg
2° to 8°C
TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members.
C13H15NO2S
Clk Inhibitor; TG003
Soluble in DMSO
Off-white
Solid
300801-52-9
249.33g/mol
Light Sensitive
Signal Transduction Reagent