Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC20 Mouse anti-Human, Clone: CDC20/1102, Abnova™

Mouse monoclonal antibody raised against full length recombinant human CDC20.

Manufacturer:  Abnova Corporation MAB13220

Catalog No. 89-933-904


Add to cart

Description

Description

CDC20 appears to act as a regulatory protein interacting with several other proteins at multiple points in the cell cycle. It is required for two microtubule-dependent processes, nuclear movement prior to anaphase and chromosome separation. [provided by RefSeq]

Specifications

Specifications

CDC20
CDC20/1102
Mouse monoclonal antibody raised against full length recombinant human CDC20.
In 10mM PBS.
CDC20A/MGC102824/bA276H19.3/p55CDC
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CDC20
Recombinant protein corresponding to full length of human CDC20.
Protein A/G purification
RUO
Primary
991
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit