Learn More
Cdc23 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP15502220UL
 View more versions of this product
Catalog No. NBP15502220
Add to cart
Description
Description
Cdc23 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications
Specifications
Cdc23
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
CDC23
Synthetic peptides corresponding to CDC23(cell division cycle 23 homolog (S. cerevisiae)) The peptide sequence was selected from the N terminal of CDC23. Peptide sequence RAAHFLHGCNSKKAYFLYMYSRYLSGEKKKDDETVDSLGPLEKGQVKNEA.
66 kDa
20ul
Cell Cycle and Replication
Primary
8697
Western Blot
Western Blot 1.25 ug/ml
Q9UJX2
ANAPC8anaphase promoting complex subunit 8, Anaphase-promoting complex subunit 8, APC8CDC23 (cell division cycle 23, yeast, homolog), cell division cycle 23 homolog (S. cerevisiae), cell division cycle protein 23 homolog, CUT23, Cyclosome subunit 8
Rabbit
IgG
Protein A purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human