Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC25B Rabbit anti-Human, Clone: JJ091-2, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267893

Catalog No. NBP267893


Add to cart

Description

Description

CDC25B Monoclonal antibody specifically detects CDC25B in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

CDC25B
JJ091-2
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
CDC25HU2, cell division cycle 25 homolog B (S. cerevisiae), cell division cycle 25 homolog B (S. pombe), cell division cycle 25B, Dual specificity phosphatase Cdc25B, EC 3.1.3.48, M-phase inducer phosphatase 2
Rabbit
IgG
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 1:100-1:2000, Immunoprecipitation
Purified
CDC25B
C terminal human Cdc25B
Protein A purified
Cancer, Cell Cycle and Replication, Phospho Specific
Primary
994
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit