Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen CDC2L1
Applications Immunofluorescence, Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092949
View Documents
Novus Biologicals
NBP303256100UL
100 μg Each for $320.71
Add to cart
 
NB092950
View Documents
Novus Biologicals
NBP30325620UL
20 μg Each for $138.37
Add to cart
 
Description

Description

CDC2L1 Polyclonal antibody specifically detects CDC2L1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence
Specifications

Specifications

CDC2L1
Unconjugated
Rabbit
Apoptosis, Cell Cycle and Replication, Cellular Markers, Protein Phosphatase, Signal Transduction
PBS with 50% glycerol, pH7.3.
984
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Immunohistochemistry
Purified
RUO
Human, Mouse, Rat
CDC2L1p58 CLK-1, CDC-related protein kinase p58, CDK11-p110, CDK11-p46, CDK11-p58, CDK11PITSLRE serine/threonine-protein kinase CDC2L1, cell division cycle 2-like 1 (PITSLRE proteins), Cell division cycle 2-like protein kinase 1, Cell division protein kinase 11B, cell division protein kinase 11B-like, CLK-1, cyclin-dependent kinase 11B, EC 2.7.11, EC 2.7.11.22, FLJ59152, Galactosyltransferase-associated protein kinase p58/GTA, p58, p58CDC2L1, p58CLK-1, PITSLREA, PK58
Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human CDC2L1 (NP_277021.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit