Learn More
Abnova™ CDC2L5 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Manufacturer:  Abnova™ L033T6
Catalog No. 89-938-877
Add to cart
Description
Description
Specifications
Specifications
Immunoprecipitation, Western Blot
CDC2L5 HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
pCMV-CDC2L5 full length ;pCMV-CDC2L5 full length
100 μg
Human
HEK293
BC001274 ;BC001274
Theoretical M.W. 37kDa
Transient overexpression cell lysate was tested with anti-CDC2L5 antibody (Mfr. No. H00008621-M02) by Western Blot.
In modified RIPA Lysis Buffer. ;In modified RIPA Lysis Buffer.
2 mg/mL, 2 mg/mL