Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC37 Rabbit anti-Human, Clone: SD08-50, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267807

Catalog No. NBP267807


Add to cart

Description

Description

CDC37 Monoclonal antibody specifically detects CDC37 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

CDC37
SD08-50
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37
Rabbit
IgG
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 1:100-1:2000, Immunoprecipitation
Purified
CDC37
Synthetic phospho-peptide surrounding S13 of CDC37
Protein A purified
Cell Cycle and Replication, Membrane Trafficking and Chaperones, Phospho Specific
Primary
11140
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit