Learn More
CDC42EP1 Rabbit anti-Human, Unconjugated, Clone: IOE-3, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB19867
Catalog No. 89-336-268
Add to cart
Description
Description
CDC42EP1 Monoclonal antibody specifically detects CDC42EP1 in Human samples. It is validated for WB-Ce.
Specifications
Specifications
CDC42EP1
IOE-3
CDC42 effector protein (Rho GTPase binding) 1
Liquid
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human
Western Blot
Unconjugated
CDC42EP1
BORG5, CEP1, MGC15316, MSE55
A synthetic peptide corresponding to human CDC42EP1
100 μg
Primary
11135