Learn More
Aviva Systems Biology Corp CDC42EP4 Antibody - N-terminal region (ARP54839_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54839_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC42EP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CDC42EP4. Alternate names: BORG4, CEP4, KAIA1777, MGC17125, MGC3740ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61337