Learn More
CDC42SE2 Mouse anti-Human, Clone: OTI10A5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808200
Catalog No. 50-166-8687
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Probably involved in the organization of the actin cytoskeleton by acting downstream of CDC42, inducing actin filament assembly. Alters CDC42-induced cell shape changes. In activated T-cells, may play a role in CDC42-mediated F-actin accumulation at the immunological synapse. May play a role in early contractile events in phagocytosis in macrophages.
Specifications
Specifications
CDC42SE2
Monoclonal
Unconjugated
CDC42SE2
Lyophilized
CDC42SE2
Full length human recombit protein of human CDC42SE2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI10A5
PBS with 8% trehalose and no preservative; pH 7.3
Q9NRR3
SPEC2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
56990