Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC42SE2 Mouse anti-Human, Clone: OTI1B12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526948

Catalog No. PIMA526948


Add to cart

Description

Description

Probably involved in the organization of the actin cytoskeleton by acting downstream of CDC42, inducing actin filament assembly. Alters CDC42-induced cell shape changes. In activated T-cells, may play a role in CDC42-mediated F-actin accumulation at the immunological synapse. May play a role in early contractile events in phagocytosis in macrophages.
Specifications

Specifications

CDC42SE2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NRR3
CDC42SE2, SPEC2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56990
Immunohistochemistry (Paraffin), Western Blot
OTI1B12
Unconjugated
CDC42SE2
Liquid
CDC42SE2
Full length human recombit protein of CDC42SE2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit