Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CDC47/MCM7 Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Specifications

Antigen CDC47/MCM7 Ab-2
Clone 47DC141; same as DCS-141
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS862R1
View Documents
Thermo Scientific™
MS862R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in Immunohistochemistry, Western Blotting, Immunofluorescence and Immunoprecipitation experiments with Thermo Scientific™ CDC47/MCM7 Ab-2, Mouse Monoclonal Antibody.

hCDC47 is a human member of the MCM family, which allows DNA to replicate once per cell cycle. In quiescent cells, human MCM7 (hMCM7) mRNA is almost undetectable. Stimulation of cells to enter the cell cycle results in induction of hMCM7 expression. The hCDC47 protein expression and localization is found in nuclei of the proliferative components of normal lymph nodes, bone marrow, epidermis and mucosa. Malignant tumors from several organs contained more nuclear hCDC47-positive cells than their normal counterparts. These results indicate that hCDC47 may play a role in normal and neoplastic cell growth in vivo.

Host Species: Mouse

Clone: 47DC141; same as DCS-141

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Dog and Xenopus. Others not known.

Epitope: Not determined

Immunogen: Recombinant hCDC47 protein

Molecular Weight: 80 kDa

Positive Control: MAD109, or PC12 cells, Tonsil or breast cancer

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CDC47/MCM7 Ab-2
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Cell Cycle
Recombinant hCDC47 protein
47DC141; same as DCS-141
Monoclonal
RUO
Primary
IgG1
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit