Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc5L Rabbit anti-Human, Mouse, Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Manufacturer:  Invitrogen 711866

Encompass

Catalog No. 71-186-6


Add to cart

Description

Description

This antibody is predicted to react with Monkey, Rabbit, Cat, Horse. Recombit rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombit monoclonal antibodies, providing the best of both worlds – the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombit antibody. While functionally the same as a polyclonal antibody – recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets – a recombit rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

CDC5L is a DNA-binding protein involved in cell cycle control. It is a component of the PRP19-CDC5L complex that forms an integral part of the spliceosome and is required for activating pre-mRNA splicing. The PRP19-CDC5L complex may also play a role in the response to DNA damage. A chromosomal aberration involving CDC5L is found in multicystic renal dysplasia.
Specifications

Specifications

Cdc5L
Recombinant Polyclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q99459 , Q6A068
KIAA0432, PCDC5RP
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
71702, 988
Western Blot
22HCLC
Unconjugated
CDC5L
Liquid
CDC5L
Protein corresponding to human Cdc5L [aa1-aa200]
Protein A
RUO
Antibody
Oligoclonal
Human, Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit