Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdc6 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297296PCP
Description
Cdc6 Polyclonal antibody specifically detects Cdc6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cdc6 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
CDC18LHsCDC18, Cdc18-related protein, CDC6 (cell division cycle 6, S. cerevisiae) homolog, CDC6 cell division cycle 6 homolog, CDC6 cell division cycle 6 homolog (S. cerevisiae), CDC6-related protein, cell division control protein 6 homolog, cell division cycle 6 homolog (S. cerevisiae), cell division cycle 6 protein, HsCdc18, HsCDC6, p62(cdc6) | |
Produced in rabbits immunized with E. coli-derived Human Cdc6 fragment. | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
990 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction