Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc6 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI1A4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270383AF405

Catalog No. N270383A405


Add to cart

Description

Description

Cdc6 Monoclonal antibody specifically detects Cdc6 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Cdc6
OTI1A4
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CDC6
Full length recombinant protein of human Cdc6 (NP_001245) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication, Core ESC Like Genes, Phospho Specific, Stem Cell Markers
Primary
990.0
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CDC18LHsCDC18, Cdc18-related protein, CDC6 (cell division cycle 6, S. cerevisiae) homolog, CDC6 cell division cycle 6 homolog, CDC6 cell division cycle 6 homolog (S. cerevisiae), CDC6-related protein, cell division control protein 6 homolog, cell division cycle 6 homolog (S. cerevisiae), cell division cycle 6 protein, HsCdc18, HsCDC6, p62(cdc6)
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit