Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CDC7 Kinase Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Specifications

Antigen CDC7 Kinase Ab-1
Clone DCS-341
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1888R1
View Documents
Thermo Scientific™
MS1888R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ CDC7 Kinase Ab-1, Mouse Monoclonal Antibody.

CDC7, a catalytic subunit of Dbf4 is an important regulator of S-phase in human cells.

Host Species: Mouse

Clone: DCS-341

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: GST fusion protein full length CDC7

Positive Control: HeLa cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CDC7 Kinase Ab-1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Primary
IgG1
DCS-341
Monoclonal
Murine
Cell Cycle
GST fusion protein full length CDC7
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit