Learn More
CDC73/HRPT2 Rabbit anti-Drosophila, Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259257
Catalog No. NBP259257
Add to cart
Description
Description
CDC73/HRPT2 Polyclonal antibody specifically detects CDC73/HRPT2 in Human, Drosophila samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.
Specifications
Specifications
CDC73/HRPT2
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
C1orf28hyperparathyroidism 2 (with jaw tumor), cell division cycle 73, Paf1/RNA polymerase II complex component, homolog (S.cerevisiae), Cell division cycle protein 73 homolog, chromosome 1 open reading frame 28, HPTJT, HRPT2FLJ23316, Hyperparathyroidism 2 protein, HYX, parafibromin
Rabbit
IgG
50 ug
Cancer, Tumor Suppressors
Primary
79577
Western Blot, ChIP assay, Immunoprecipitation
Western Blot 12ug, Chromatin Immunoprecipitation 1:1000, Immunoprecipitation 1-5 ug
Purified
CDC73
The exact immunogen is propietary information.
Protein G purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Drosophila