Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC73 Mouse anti-Human, Clone: 2H1-F10-B9, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | CDC73 |
---|---|
Clone | 2H1-F10-B9 |
Concentration | 1.0 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC176825
|
MilliporeSigma
MABC176825UG |
25 μg |
Each for $177.00
|
|
|||||
MABC1768100
|
MilliporeSigma
MABC1768100UG |
100 μg |
Each for $450.00
|
|
|||||
Description
CDC73 Monoclonal antibody specifically detects CDC73 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.Specifications
CDC73 | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG1 κ | |
Protein G purified |
2H1-F10-B9 | |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
KLH-conjugated linear peptide corresponding to 13 amino acids from the N-terminal region of human CDC73 (Parafibromin). | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title