Learn More
Aviva Systems Biology Corp CDCA4 Antibody - N-terminal region (ARP50622_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50622_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CDCA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CDCA4. Alternate names: FLJ20764, HEPP, MGC19517, SEI-3/HEPPApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-60391