Learn More
CDCA8 293T Cell Overexpression Lysate (Denatured), Gene Accession Number: NM_018101.2, Abnova
Manufacturer:  Abnova Corporation H00055143T02
Catalog No. 89-002-414
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_018101.2
6.8
Transient overexpression cell Lysate was tested with Anti-CDCA8 antibody (H00055143-B02) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
CDCA8 293T Cell Transient Overexpression Lysate(Denatured)
31.3
pCMV-CDCA8 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.