Learn More
Aviva Systems Biology Corp CDH1 Antibody - middle region (ARP51289_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51289_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CDH1. Alternate names: Arc-1, CD324, CDHE, ECAD, LCAM, UVOApplicationIF, WB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60537