Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-cadherin Mouse anti-Human, APC, Clone: 67A4, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A15717

Catalog No. 50-113-7733


Add to cart

Description

Description

E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintece of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.
Specifications

Specifications

E-cadherin
Monoclonal
APC
CDH1
Liquid
CDH1
Human CD324
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
67A4
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P12830
Arc-1, CD324, CDHE, ECAD, LCAM, UVO, CAM 120/80, E-Cadherin, cadherin 1, E-cadherin epithelial, cadherin-1, calcium-dependent adhesion protein, epithelial, cell-CAM 120/80, epithelial cadherin, uvomorulin, cadherin-E
Mouse
IgG1
100 tests
4° C
Primary
999
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit