Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDH17 Mouse anti-Human, Unconjugated, Clone: Lic5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT152525UG
Description
CDH17 Monoclonal antibody specifically detects CDH17 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition, Western Blotting.Specifications
CDH17 | |
Monoclonal | |
Unconjugated | |
Intestinal peptide-associated transporter HPT-1;Liver-intestine cadherin;LI-cadherin | |
Mouse | |
92.22 kDa calculated. | |
25 μg | |
Primary | |
Human | |
Purified |
Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot | |
Lic5 | |
NP_001138135 | |
CDH17 | |
Recombinant fragment corresponding to extracellular domains 1 and 2 of human Cadherin-17 (CDH17). | |
Protein G Purified | |
Cell Structure | |
Clone Lic5 is a mouse monoclonal antibody that specifically detects human Cadherin-17. It targets an epitope within the extracellular domain. | |
Stable for 1 year at -20°C from date of receipt. | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction