Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDH18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536240
Description
This antibody detects endogenous protein at a molecular weight of 88 kDa. Purity is >95% by SDS-PAGE.
This gene encodes a type II classical cadherin from the cadherin superfamily of integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is expressed specifically in the central nervous system and is putatively involved in synaptic adhesion, axon outgrowth and guidance. Cadherins are calcium dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types.
Specifications
CDH18 | |
Polyclonal | |
Unconjugated | |
CDH18 | |
B230220E17Rik; Cadherin; Cadherin 14; cadherin 18; cadherin 18, type 2; Cadherin14; cadherin-14; Cadherin18; Cadherin-18; cadherin-like 24; CDH14; Cdh14l; CDH18; CDH24; ey-cadherin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1016, 310174, 320865 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.2 | |
Q13634 | |
CDH18 | |
Synthetic peptide corresponding to amino acids 96-140 of human Cadherin-18. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction