Learn More
Abnova™ CDH19 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00028513T01
Catalog No. 89-938-975
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CDH19 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 85.03kDa
Transient overexpression cell lysate was tested with anti-CDH19 antibody (Mfr. No. H00028513-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_021153.2
pCMV-CDH19 full-length
100 μL
Human