Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH19 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548480

Catalog No. PA548480

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types.
Specifications

Specifications

CDH19
Polyclonal
PBS with 0.09% sodium azide
Q9H159
Cadherin-19, CDH19, CDH7L2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
28513
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CDH19
Liquid
CDH19
KLH conjugated synthetic peptide between 613-640 amino acids from the C-terminal region of human CDH19.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit