Learn More
CDH19 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA550923
Catalog No. PA550923
Add to cart
Description
Description
CDH19 Polyclonal Antibody detects endogenous levels of CDH19 protein.
This gene is one of three related type II cadherin genes situated in a cluster on chromosome 18. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein containing five extracellular cadherin repeats. Loss of cadherins may be associated with cancer formation. Alternative splicing results in multiple transcript variants for this gene. [provided by RefSeq, Aug 2012].
Specifications
Specifications
CDH19
Polyclonal
Unconjugated
CDH19
Liquid
CDH19
Synthesized peptide derived from the Internal region of human Cadherin-19.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9H159
CDH7L2
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
28513