Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

CDH2 Mouse anti-Canine, Human, Mouse, Clone: OTI2G7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503933

Catalog No. 50-167-4710



Description

Description

CDH2 encodes a classical cadherin and member of the cadherin superfamily. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein is proteolytically processed to generate a calcium-dependent cell adhesion molecule and glycoprotein. This protein plays a role in the establishment of left-right asymmetry, development of the nervous system and the formation of cartilage and bone.
Specifications

Specifications

CDH2
Monoclonal
0.77 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P15116, P19022
CD325, CDHN, CDw325, NCAD
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1000, 12558, 480169
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2G7
Unconjugated
CDH2
Liquid
CDH2
Full length human recombit protein of human CDH2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit