Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH2 Mouse anti-Human, Mouse, Clone: CDH2/1573, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14462

Catalog No. 89-034-545


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human CDH2.

  • High specificity
  • High reactivity
Specifications

Specifications

CDH2
CDH2/1573
Mouse monoclonal antibody raised against partial recombinant human CDH2.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
CDH2
Recombinant protein corresponding to intracellular domain of human CDH2.
Protein A/G purification
RUO
Primary
1000
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined b
P19022
CD325|CDHN|CDw325|NCAD
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)