Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDH7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA511540
Description
This antibody is predicted to react with chicken and mouse based on sequence homology.
This gene is a type II classical cadherin from the cadherin superfamily. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Cadherins mediate cell-cell binding in a homophilic manner, contributing to the sorting of heterogeneous cell types and the maintenance of orderly structures. Alternative splicing in the 5' UTR of this gene yields variant transcripts encoding the same protein.
Specifications
CDH7 | |
Polyclonal | |
Unconjugated | |
CDH7 | |
9330156F07Rik; Cad7; Cadherin; cadherin 7; cadherin 7, type 2; Cadherin7; cadherin-7; Cdh7; CDH7L1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1005 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9ULB5 | |
CDH7 | |
KLH conjugated synthetic peptide between 56-84 amino acids from the N-terminal region of human CDH7. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction