Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDHR4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551317

Catalog No. PA551317


Add to cart

Description

Description

The antibody detects endogenous levels of total CDHR4 protein.

Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types .
Specifications

Specifications

CDHR4
Polyclonal
Unconjugated
CDHR4
Liquid
CDHR4
Fusion protein of human CDHR4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.5 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
A6H8M9
CDH29
Rabbit
IgG
100 μL
-20°C
Primary
389118
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit