Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cdk1/p34 cdc2 Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and kinase assay applications. 

Specifications

Antigen Cdk1/p34superscriptcdc2superscript Ab-1
Clone A17.1.1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS110R1
View Documents
Thermo Scientific™
MS110R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and kinase assay applications with Thermo Scientific™ Cdk1/p34cdc2 Ab-1, Mouse Monoclonal Antibody.

p34cdc2(a catalytic subunit of Maturation Promoting Factor) plays a crucial role during cell division and is most active during mitosis. It is a serine/threonine kinase, which is activated by cyclin through dephosphorylation of tyrosine residues. p34cdc2 is inactivated by a tyrosine kinase.

Host Species: Mouse

Clone: A17.1.1

Isotype: IgG2a

Species Reactivity: Human, Mouse, Rat, Guinea Pig, Woodchuck, Xenopus and Chicken.

Epitope: aa 220-227

Immunogen: C-Terminal 2/3 of Xenopus CDC2 expressed in E. coli

Molecular Weight: 34 kDa

Positive Control: HeLa, MAD109, PC12 or NIH-3T3 cells; Tonsil or lymph node

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Kinase Assay
  • Inhibits activation of p34cdc2 kinase by cyclins

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cdk1/p34superscriptcdc2superscript Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Cell Cycle
C-Terminal 2/3 of Xenopus CDC2 expressed in E. coli
A17.1.1
Monoclonal
RUO
Primary
IgG2a
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit