Learn More
CDK10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 171821AP
Catalog No. 50-173-1426
Add to cart
Description
Description
The protein encoded by this gene belongs to the CDK subfamily of the Ser/Thr protein kinase family. The CDK subfamily members are highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2, and are known to be essential for cell cycle progression. This kinase has been shown to play a role in cellular proliferation and its function is limited to cell cycle G2-M phase. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG (CUG) translation initiation codon, have been found for this gene.
Specifications
Specifications
CDK10
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15131, Q3UMM4, Q4KM47
CDK10, cyclin dependent kinase 10, PISSLRE
Rabbit
IgG
150 μL
-20°C
Primary
234854, 361434, 8558
Western Blot
Unconjugated
CDK10
Liquid
Cdk10
CDK10 Fusion Protein Ag10935
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat