Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFTK2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548681

Catalog No. PA548681

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Serine/threonine-protein kinase involved in the control of the eukaryotic cell cycle, whose activity is controlled by an associated cyclin.
Specifications

Specifications

PFTK2
Polyclonal
PBS with 0.09% sodium azide
Q96Q40, Q3V3A1
Cyclin-dependent kinase 15, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 7 protein homolog, Cell division protein kinase 15, Serine/threonine-protein kinase ALS2CR7, Serine/threonine-protein kinase PFTAIRE-2, Cdk15, Als2cr7, Pftk2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
271697, 65061
Western Blot
Unconjugated
CDK15
Liquid
CDK15
KLH conjugated synthetic peptide between 96-124 amino acids from the N-terminal region of mouse Cdk15.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit