Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCTAIRE2 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548702

Catalog No. PA548702

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May play a role in terminally differentiated neurons. Has a Ser/Thr-phosphorylating activity for histone H1.
Specifications

Specifications

PCTAIRE2
Polyclonal
PBS with 0.09% sodium azide
Q8K0D0
Cyclin-dependent kinase 17, Cell division protein kinase 17, PCTAIRE-motif protein kinase 2, Serine/threonine-protein kinase PCTAIRE-2, Cdk17, Pctaire2, Pctk2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
237459
Western Blot
Unconjugated
Cdk17
Liquid
CDK17
KLH conjugated synthetic peptide between 484-513 amino acids from the C-terminal region of mouse Cdk17.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit