Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCTAIRE3 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548730

Catalog No. PA548730

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with rat based on sequence homology.

May play a role in signal transduction cascades in terminally differentiated cells.
Specifications

Specifications

PCTAIRE3
Polyclonal
PBS with 0.09% sodium azide
Q04899
Cyclin-dependent kinase 18, Cell division protein kinase 18, PCTAIRE-motif protein kinase 3, Serine/threonine-protein kinase PCTAIRE-3, Cdk18, Pctaire3, Pctk3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18557
Western Blot
Unconjugated
Cdk18
Liquid
CDK18
KLH conjugated synthetic peptide between 422-449 amino acids from the C-terminal region of mouse Cdk18.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit