Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cdk2 Ab-1, Mouse Monoclonal Antibody

Accurately detect Cdk2 in human, mouse and rat tissues with Thermo Scientific™ Cdk2 Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen Cdk2 Ab-1
Clone 2B6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS459R1
View Documents
Thermo Scientific™
MS459R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Accurately detect Cdk2 in human, mouse and rat tissues with Thermo Scientific™ Cdk2 Ab-1, Mouse Monoclonal Antibody.

p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as is cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.

Host Species: Mouse

Clone: 2B6

Isotype: IgG2b

Species Reactivity: Human, Mouse and Rat.

Epitope: Not determined

Immunogen: Human recombinant Cdk2 protein

Molecular Weight: 33 kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cdk2 Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Murine
Cell Cycle
Human recombinant cdk2 protein
2B6
Monoclonal
RUO
Primary
IgG2b
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit