Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK2/Cyclin A Recombinant Human Protein Non-distribution product as customer accommodation.

CDK2/Cyclin A Recombinant Human Protein

$432.00 - $6290.00

Specifications

Accession Number NP_001789, NP_001228
Assays CDK2/cyclin A was pre-diluted in enzyme dilution buffer and assayed in 20mM Tris (pH 7.5), 5mM MgCl2, 100μM ATP 100μM histone H1 peptide substrate (PKTPKKAKKL) and trace [32P]-γ-ATP for 10 minutes at 30°C
Common Name CDK2/Cyclin A Recombinant Human Protein
Concentration 0.17mg/mL
Expression System Baculovirus
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3267 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV3267
10μg Each for $432.00
Add to cart
 
PR4448A Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PR4448A
1mg Each for $6,290.00
Add to cart
 
Description

Description

CDK2 is a catalytic subunit of the cyclin-dependent protein kinase complex, and is essential for cell cycle G1⁄S phase transition. This protein is coexpressed and copurified with CyclinA.
Specifications

Specifications

NP_001789, NP_001228
CDK2/Cyclin A Recombinant Human Protein
Baculovirus
CDK and CHK
1 Each
90% by SDS-PAGE
4,560nmoles of phosphate transferred to histone H1 peptide substrate (PKTPKKAKKL) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.5μg/mL
Kinase assay
His Tag
CDK2/cyclin A was pre-diluted in enzyme dilution buffer and assayed in 20mM Tris (pH 7.5), 5mM MgCl2, 100μM ATP 100μM histone H1 peptide substrate (PKTPKKAKKL) and trace [32P]-γ-ATP for 10 minutes at 30°C
0.17mg/mL
CDK2/cyclin A
CDK2/Cyclin A Recombinant Human Protein
CDK2/Cyclin A Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
–80°C
Human

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit