Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDK2AP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104487
Description
Antibody detects endogenous levels of total CDKA2.
This gene encodes a protein that interacts with cyclin-dependent kinase 2 associated protein 1. Pseudogenes associated with this gene are located on chromosomes 7 and 9. Alternatively spliced transcript variants have been observed for this gene.
Specifications
CDK2AP2 | |
Polyclonal | |
Unconjugated | |
CDK2AP2 | |
5830466O21Rik; Cdk2ap2; CDK2-associated protein 2; cyclin dependent kinase 2 associated protein 2; cyclin-dependent kinase 2 associated protein 2; cyclin-dependent kinase 2-associated protein 2; D19Ertd144e; DOC-1 related protein; Doc1r; DOC-1R; DOC-1-related protein; p14; tumor suppressor deleted in oral cancer related 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10263, 52004 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75956, Q9CPY4 | |
CDK2AP2 | |
A synthesized peptide derived from human CDKA2, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction