Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cdk4 Ab-1, Mouse Monoclonal Antibody

Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications.

$173.10 - $948.00

Specifications

Antigen Cdk4 Ab-1
Clone DCS-35
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Format Concentrated
View More Specs

Products 8
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity & Availability  
MS299B
View Documents
Thermo Scientific™
MS299B
200μg/mL Biotin 1mL Each for $948.00
Add to cart
 
MS299B0
View Documents
Thermo Scientific™
MS299B0
200μg/mL Biotin 100μL Each for $232.00
Add to cart
 
MS299B1
View Documents
Thermo Scientific™
MS299B1
200μg/mL Biotin 500μL Each for $635.50
Add to cart
 
MS299P
View Documents
Thermo Scientific™
MS299P
200μg/mL Unconjugated 1mL Each for $774.50
Add to cart
 
MS299P0
View Documents
Thermo Scientific™
MS299P0
200μg/mL Unconjugated 100μL Each for $173.10
Add to cart
 
MS299P1
View Documents
Thermo Scientific™
MS299P1
200μg/mL Unconjugated 500μL Each for $482.50
Add to cart
 
MS299P1ABX
View Documents
Thermo Scientific™
MS299P1ABX
1mg/mL Unconjugated 100μL Each for $537.00
Add to cart
 
MS299PABX
View Documents
Thermo Scientific™
MS299PABX
1mg/mL Unconjugated 200μL Each for $798.00
Add to cart
 
Description

Description

Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications with Thermo Scientific™ Cdk4 Ab-1, Mouse Monoclonal Antibody.

A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: DCS-35

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat and Pig. Others not known.

Epitope: aa 1-20

Immunogen: Purified recombinant Cdk4 protein

Molecular Weight: 34 kDa

Positive Control: NIH 3T3, MAD109, LS174T, or MCF-7 cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cdk4 Ab-1
Immunofluorescence, Immunoprecipitation, Western Blot
Concentrated
RUO
Primary
IgG2a kappa
DCS-35
Monoclonal
Murine
Cell Cycle
Purified recombinant cdk4 protein
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit