Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cdk4 Ab-6, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk4 Ab-6, Mouse Monoclonal Antibody.

Specifications

Antigen Cdk4 Ab-6
Clone DCS-31 + DCS-35
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
MS616P0
View Documents
Thermo Scientific™
MS616P0
200μg/mL 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS616P1
View Documents
Thermo Scientific™
MS616P1
200μg/mL 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cdk4 Ab-6, Mouse Monoclonal Antibody.

A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: DCS-31 + DCS-35

Isotype: IgG1 + IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 1-35

Immunogen: Purified recombinant Cdk4 protein

Molecular Weight: 34 kDa

Positive Control: MCF-7 or PC12 cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cdk4 Ab-6
Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Murine
Cell Cycle
Purified recombinant cdk4 protein
DCS-31 + DCS-35
Monoclonal
Concentrated
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit