Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK4 Mouse anti-Human, Alexa Fluor™ 594, Clone: 6D10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242642AF594
Description
CDK4 Monoclonal specifically detects CDK4 in Human samples. It is validated for Western Blot, ELISA.Specifications
CDK4 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CDK4 | |
Recombinant human CDK4 (1-303aa) purified from E. coli | |
Primary | |
Human | |
IgG2a κ |
Western Blot, ELISA | |
6D10 | |
Western Blot, ELISA | |
Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4 | |
Mouse | |
100 μL | |
1019 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title