Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk5 Mouse anti-Human, Canine, Janelia Fluor™ 549, Clone: OTI2H6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270396JF549

Catalog No. N270396J549


Add to cart

Description

Description

Cdk5 Monoclonal antibody specifically detects Cdk5 in Human, Canine samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Cdk5
OTI2H6
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
CDK5
Full length recombinant protein of human Cdk5 (NP_004926) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication, Innate Immunity, Phospho Specific
Primary
1020.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit